Anti-COL4A5 Rabbit Polyclonal Antibody
Catalog # 76749-364
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:collagen type IV alpha 5 chain
- Antigen Symbol:COL4A5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:1287
- Antigen Synonyms:ATS1|dA149D17.3|collagen of basement membrane|ATS|ASLN|collagen alpha-5(IV) chain|collagen|alpha 5|alpha-5 chain|type IV|collagen IV|CA54|dA24A23.1|alpha-5 polypeptide
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1462-1691 of human COL4A5 (NP_203699.1).
- Purification:Affinity purification.
- Cat. No.:76749-364
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: COL4A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human