Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-MAT2A Rabbit Polyclonal Antibody
Catalog # 76755-142
Supplier:  Prosci
Anti-MAT2A Rabbit Polyclonal Antibody
Catalog # 76755-142
Supplier:  Prosci
Supplier Number:  23-582

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    methionine adenosyltransferase II, alpha
  • Antigen Symbol:
    MAT2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    50 µl
  • Form:
    Liquid
  • Gene ID:
    4144
  • Antigen Synonyms:
    SAMS2|MAT 2|MAT2A|MAT-II|AMS2|MATA2|MATII|AdoMet synthetase 2|AdoMet synthase 2
  • Modification:
    None
  • Storage Buffer:
    PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
  • Storage Temperature:
    Store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping Temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-395 of human MAT2A (NP_005902.1).
  • Purification:
    Affinity purification.
  • Cat. No.:
    76755-142
  • Packaging:
    Vial

Specifications

About this item

Type: Primary
Antigen: MAT2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat