Nprl3 Peptide
Vial, 0.1 mg
77128-314
: Prosci
: 42-928P
- Pack type:Vial
- Size:0.1 mg
- Storage Conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Cat. No.:77128-314
- Shipping Temperature:RT
- Protein/Peptide Type:Synthetic
- Gene ID:17168
- Formulation:Lyophilized. Nprl3 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Tested Applications:Blocking
- Protein Synonyms:Mare|HS-40|CGTHBA|Alpha-globin regulatory element-containing gene protein|Nitrogen permease regulator 3-like protein|Prox1|Phg|m(alpha)RE|Aag|HS-26
- Protein/Peptide Name:Nprl3
- Conjugation:Unconjugated
- Species:Human
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



