Mouse Recombinant LAIR1/CD305 Protein (from HEK293 cells)
77119-368
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:0.1 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:52855
- Protein Synonyms:LAIR1|CD305
- Protein/Peptide Name:Lair1
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:40.6 kDa
- Sequence:Gln 22 - Tyr 141
- Formulation:Lyophilized. 50 mM Tris, 100 mM Glycine, pH 7.5.
- Shipping Temperature:RT
- Tested Applications:ELISA,WB
- Cat. No.:77119-368