Human Recombinant GITR/TNFRSF18 Protein (from HEK293 cells)
Vial, 0.1 mg
Catalog # 77128-620
Supplier: Prosci
Supplier #: 11-486
Human Recombinant GITR/TNFRSF18 Protein (from HEK293 cells)
Catalog # 77128-620
Supplier: Prosci
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Specifications
Specifications
- Pack type:Vial
- Size:0.1 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Molecular Weight:41.0 kDa
- Purity:>95% as determined by SDS-PAGE
- Tested Applications:ELISA,WB
- Sequence:Gln 26 - Glu 161
- Source:HEK293 cells
- Cat. No.:77128-620
- Shipping Temperature:RT
- Protein/Peptide Type:Recombinant
- Gene ID:8784
- Formulation:Lyophilized. Tris with Glycine, Arginine and NaCl, pH 7.5.
- Protein Synonyms:CD357|GITR|TNFRSF18|AITR
- Protein/Peptide Name:TNFRSF18
- Conjugation:Unconjugated
- Species:Human
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



