Mouse Recombinant CD27 Ligand/CD70 Recombinant Protein (from HEK293 cells)
Catalog # 77121-822
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:0.1 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:21948
- Protein Synonyms:CD27LG|TNFSF7|TNFSF7G|CD27L|CD70
- Protein/Peptide Name:Cd70
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:18.6 kDa
- Sequence:Gln 47 - Pro 195
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping Temperature:RT
- Tested Applications:ELISA,WB
- Cat. No.:77121-822