Human Recombinant NTB-A/SLAMF6 Protein (from HEK293 cells)
77128-624
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.1 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:114836
- Protein Synonyms:SLAMF6|NK-T-B-antigen|CD352|KALI|NTB-A|Ly108
- Protein/Peptide Name:SLAMF6
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:25.0 kDa
- Sequence:Gln 22 - Met 226
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping Temperature:RT
- Tested Applications:WB
- Cat. No.:77128-624