Human Recombinant IL-1 Rrp2/IL-1 R6 Protein (from HEK293 cells)
77126-060
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.1 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:8808
- Protein Synonyms:IL1RL2|IL-1RL2|IL-36R|IL-1Rrp2|IL1Rrp2|IL-1 R6|IL1RRP2|IL-1 Rrp2
- Protein/Peptide Name:IL1RL2
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:37.8 kDa
- Sequence:Asp 20 - Arg 335
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping Temperature:RT
- Tested Applications:WB
- Cat. No.:77126-060