Rhesus monkey Recombinant CD155/PVR Protein (from HEK293 cells)
Catalog # 77124-724
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rhesus monkey
- Size:0.1 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:712851
- Protein Synonyms:TAGE4|PVS|FLJ25946|CD155|NECL5|PVR|HVED
- Protein/Peptide Name:PVR
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:36.3 kDa
- Sequence:Asp 28 - Asn 343
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping Temperature:RT
- Tested Applications:ELISA,WB
- Cat. No.:77124-724