Human Recombinant IL-23 R Protein (from HEK293 cells)
77119-448
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.1 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:149233
- Protein Synonyms:IL-23 R|IL-23 Receptor
- Protein/Peptide Name:IL23R
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:64.5 kDa
- Sequence:Gly 24 - Gly 355
- Formulation:Lyophilized. Tris with Glycine, Arginine and NaCl, pH 7.5.
- Shipping Temperature:RT
- Tested Applications:ELISA,WB
- Cat. No.:77119-448