Human Recombinant Siglec-8 Protein (from HEK293 cells)
77131-060
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.1 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:2817
- Protein Synonyms:SIGLEC8|SAF2|SAF-2|Siglec-8
- Protein/Peptide Name:GPC1
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:39.7 kDa
- Sequence:Met 17 - Ala 363
- Formulation:Lyophilized. PBS with Arginine, pH 7.4.
- Shipping Temperature:RT
- Tested Applications:WB
- Cat. No.:77131-060