Human Recombinant Frizzled-7/FZD7 Protein (from HEK293 cells)
Catalog # 77123-888
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.1 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:8324
- Protein Synonyms:Fz-7|FzE3|hFz7|Frizzled-7|FZD7
- Protein/Peptide Name:FZD7
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:18.6 kDa
- Sequence:Gln 33 - Leu 185
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping Temperature:RT
- Tested Applications:WB
- Cat. No.:77123-888