Cynomolgus monkey Recombinant Siglec-9 Protein (from HEK293 cells)
Catalog # 77130-868
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Size:0.1 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:102118310
- Protein Synonyms:SIGLEC9|Siglec-9|CDw329|CD329
- Protein/Peptide Name:SIGLEC9
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:38.0 kDa
- Sequence:Gln 20 - Gly 350
- Formulation:Lyophilized. 25 mM MES, 150 mM NaCl, pH 5.5.
- Shipping Temperature:RT
- Tested Applications:ELISA,WB
- Cat. No.:77130-868