Human Recombinant Biotinylated PSGL-1/CD162 Protein (from HEK293 cells)
Catalog # 77119-500
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.025 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:6404
- Protein Synonyms:CD162|Selectin P ligand|PSGL1|SELPLG
- Protein/Peptide Name:SELPLG
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:32.4 kDa
- Sequence:Gln 42 - Cys 320
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping Temperature:RT
- Tested Applications:WB
- Cat. No.:77119-500