Order Entry
Puerto Rico
ContactUsLinkComponent
SARS-CoV-2 (COVID-19) Recombinant Trimeric Spike (S) Protein (from HEK293 cells)
SARS-CoV-2 (COVID-19) Recombinant Trimeric Spike (S) Protein (from HEK293 cells)
Catalog # 77120-146
Supplier:  Prosci
CAS Number:  
SARS-CoV-2 (COVID-19) Recombinant Trimeric Spike (S) Protein (from HEK293 cells)
Catalog # 77120-146
Supplier:  Prosci
Supplier Number:  10-075
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    SARS-CoV-2
  • Size:
    0.1 mg
  • Storage Conditions:
    This recombinant protein may be stored as received at 2...8 °C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at −80 °C. Avoid Repeated Freeze Thaw Cycles.
  • Gene ID:
    1791269090
  • Protein/Peptide Name:
    Trimeric Spike (S)
  • Purity:
    >95% by SDS PAGE Endotoxin level ≤1.0 EU/mg as determined by the LAL method
  • Molecular Weight:
    The predicted molecular mass is ~132 kDa for monomer and ~396 kDa for trimer
  • Formulation:
    Liquid. This recombinant protein is aseptically packaged and formulated in 0.01 M phosphate buffered saline (PBS) pH 7.2 - 7.4, 150 mM NaCl with no carrier protein, potassium, calcium or preservatives added.
  • Shipping Temperature:
    Dry Ice
  • Tested Applications:
    ELISA
  • Cat. No.:
    77120-146

Specifications

About this item

QCVNLTTRTQLPPAYTNSFTRGVYYPDKVFRSSVLHSTQDLFLPFFSNVTWFHAIHVSGTN
GTKRFDNPVLPFNDGVYFASTEKSNIIRGWIFGTTLDSKTQSLLIVNNATNVVIKVCEFQFC
NDPFLGVYYHKNNKSWMESEFRVYSSANNCTFEYVSQPFLMDLEGKQGNFKNLREFVFK
NIDGYFKIYSKHTPINLVRDLPQGFSALEPLVDLPIGINITRFQTLLALHRSYLTPGDSSSGWT
AGAAAYYVGYLQPRTFLLKYNENGTITDAVDCALDPLSETKCTLKSFTVEKGIYQTSNFRVQ
PTESIVRFPNITNLCPFGEVFNATRFASVYAWNRKRISNCVADYSVLYNSASFSTFKCYGVS
PTKLNDLCFTNVYADSFVIRGDEVRQIAPGQTGKIADYNYKLPDDFTGCVIAWNSNNLDS
KVGGNYNYLYRLFRKSNLKPFERDISTEIYQAGSTPCNGVEGFNCYFPLQSYGFQPTNGVG
YQPYRVVVLSFELLHAPATVCGPKKSTNLVKNKCVNFNFNGLTGTGVLTESNKKFLPFQQF
GRDIADTTDAVRDPQTLEILDITPCSFGGVSVITPGTNTSNQVAVLYQDVNCTEVPVAIHA
DQLTPTWRVYSTGSNVFQTRAGCLIGAEHVNNSYECDIPIGAGICASYQTQTNSPGSASS
VASQSIIAYTMSLGAENSVAYSNNSIAIPTNFTISVTTEILPVSMTKTSVDCTMYICGDSTECS
NLLLQYGSFCTQLNRALTGIAVEQDKNTQEVFAQVKQIYKTPPIKDFGGFNFSQILPDPSKP
SKRSFIEDLLFNKVTLADAGFIKQYGDCLGDIAARDLICAQKFNGLTVLPPLLTDEMIAQYTS
ALLAGTITSGWTFGAGAALQIPFAMQMAYRFNGIGVTQNVLYENQKLIANQFNSAIGKIQ
DSLSSTASALGKLQDVVNQNAQALNTLVKQLSSNFGAISSVLNDILSRLDPPEAEVQIDRLI
TGRLQSLQTYVTQQLIRAAEIRASANLAATKMSECVLGQSKRVDFCGKGYHLMSFPQSAP
HGVVFLHVTYVPAQEKNFTTAPAICHDGKAHFPREGVFVSNGTHWFVTQRNFYEPQIITT
DNTFVSGNCDVVIGIVNNTVYDPLQPELDSFKEELDKYFKNHTSPDVDLGDISGINASVVN
IQKEIDRLNEVAKNLNESLIDLQELGKYEQ