Anti-KMT5C Recombinant Antibody [Clone: RAB-C409]
Catalog # 76630-904
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:lysine methyltransferase 5C
- Antigen Symbol:KMT5C
- Clonality:Recombinant
- Clone:RAB-C409
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:84787
- Antigen Synonyms:Histone-lysine N-methyltransferase KMT5C|anti-SUV420H2-RAB-C409|Suppressor of variegation 4-20 homolog 2|Suv4-20h2|Lysine-specific methyltransferase 5C|48G1_11|2.1.1.361|Lysine N-methyltransferase 5C|2.1.1.362|Su(var)4-20 homolog 2|[histone H4]-N-methyl-L-lysine20 N-methyltransferase KMT5B|[histone H4]-lysine20 N-methyltransferase KMT5B
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:This antibody was obtained by recombinant antibody (rAb) phage display recognizing SUV420H2 protein under non-denaturing conditions.
- Cat. No.:76630-904
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: lysine methyltransferase 5C
Clonality: Recombinant
Clone: RAB-C409
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human