Anti-Vsir Recombinant Antibody [Clone: 13F3]
76629-416
: Prosci
- Antibody Type:Primary
- Antigen Name:V-set immunoregulatory receptor
- Antigen Symbol:Vsir
- Clonality:Recombinant
- Clone:13F3
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Isotype:IgG lambda
- Reactivity:Mouse
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:74048
- Antigen Synonyms:Platelet receptor Gi24|V-domain Ig suppressor of T cell activation|Vsir|V-set immunoregulatory receptor|V-type immunoglobulin domain-containing suppressor of T-cell activation|PD-1H|Dies1|V-set domain-containing immunoregulatory receptor
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising Armenian hamsters with EL4 cells overexpressing VISTA-RFP, and VISTA-Ig fusion protein.
- Tested Applications:Blocking
- Cat. No.:76629-416
- Packaging:Vial
Type: Primary
Antigen: V-set immunoregulatory receptor
Clonality: Recombinant
Clone: 13F3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG lambda
Reactivity: Mouse