Anti-MS4A4A Recombinant Antibody [Clone: 3F2]
76629-608
: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:membrane spanning 4-domains A4A
- Antigen Symbol:MS4A4A
- Clonality:Recombinant
- Clone:3F2
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:0.2 mg
- Environmentally Preferable:
- Cross Adsorption:No
- Form:Liquid
- Gene ID:51338
- Antigen Synonyms:CD20 antigen-like 1|human membrane-spanning 4-domains subfamily A member 4A|MS4A|4SPAN1|(MS4A4A)-GFP|MS4A4|Four-span transmembrane protein 1
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:This antibody was generated by immunizing C57BL/6 mice against mouse cell line 300-19 expressing transfected human membrane-spanning 4-domains subfamily A member 4A (MS4A4A)-GFP.
- Cat. No.:76629-608
- Packaging:Vial
Type: Primary
Antigen: membrane spanning 4-domains A4A
Clonality: Recombinant
Clone: 3F2
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human