Anti-L3MBTL1 Recombinant Antibody [Clone: RAB-C213]
Catalog # 76629-266
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:L3MBTL histone methyl-lysine binding protein 1
- Antigen Symbol:L3MBTL1
- Clonality:Recombinant
- Clone:RAB-C213
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:Fab fragment kappa
- Reactivity:Human
- Size:0.1 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:26013
- Antigen Synonyms:H-l(3)mbt|anti-L3MBTL1-RAB-C213|L(3)mbt protein homolog|L(3)mbt-like|L3MBTL1-A003-1453|L3MBTL1|Lethal(3)malignant brain tumor-like protein 1|Lethal 3 malignant brain tumor-like protein 1|H-l(3)mbt protein
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:This antibody was obtained by recombinant antibody (rAb) phage display recognizing L3MBTL1 protein under non-denaturing conditions.
- Cat. No.:76629-266
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: L3MBTL histone methyl-lysine binding protein 1
Clonality: Recombinant
Clone: RAB-C213
Conjugation: Unconjugated
Host: Recombinant
Isotype: Fab fragment kappa
Reactivity: Human