EZBio® Titanium Round (TR) Polycarbonate Carboys
Foxx Life Sciences’ EZBio® Titanium Round (TR) Polycarbonate Carboys are molded with ultra-pure USP Class VI resins, FDA 21 CFR 177 Grade materials. They are supplied double bagged, and are available sterile or non sterile.
- Available in 5 and 10 L sizes
- Made entirely of 100% USP Class VI components
- Compatible with leakproof VersaCap® 83B caps for flexibility of interchangeable connectors
- Carboys can withstand low temperatures down to −135 °C
- Manufactured in a Class 7 cleanroom
- Autoclavable
EZBio® Titanium Round (TR) Carboys are advanced fluid handling systems developed with the end user in mind. The EZBio® TR Carboy serves as a highly versatile container suitable for all fluid management applications. These round carboys are perfect for applications that require homogeneous mixing. The shape eliminates any pockets of undissolved material that square carboys might develop due to uneven corners.
: USP Class VI components. Manufactured in an ISO 13485 facility in North America.
: Bottles are packaged in shrinked-wrapped modules.