Order Entry
Puerto Rico
ContactUsLinkComponent
2,4,6-Trimethyl-2,4,6-trivinylcyclotrisiloxane 97% (by GC)
Flammable
Irritant
2,4,6-Trimethyl-2,4,6-trivinylcyclotrisiloxane 97% (by GC)
Catalog # 77168-092
Supplier:  Ambeed
CAS Number:  
Flammable
Irritant
2,4,6-Trimethyl-2,4,6-trivinylcyclotrisiloxane 97% (by GC)
Catalog # 77168-092
Supplier:  Ambeed
Supplier Number:  A383440-5G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    5 g
  • Cat. No.:
    77168-092

Specifications

About this item

 :
 :258.50 g/mol
UN:1993
ADR:3,Ⅲ
 :GHS02
 :GHS07
CAS Number:3901-77-7
Formula:C₉H₁₈O₃Si₃
MDL Number:MFCD00023111