Boc-D-Glu(OMe)-OH·DCHA 95%
New Product
Supplier: Ambeed
About this item
Caution: for Research Use Only
Synonyms: Boc-D-Glu(Ome)-OH.DCHA
- CAS Number:76379-02-7
- Formula:C₁₁H₁₉NO₆·C₁₂H₂₃N
- MDL Number:MFCD01861340
- Molecular Weight:442.6 g/mol
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Caution: for Research Use Only
Synonyms: Boc-D-Glu(Ome)-OH.DCHA
- CAS Number:76379-02-7
- Formula:C₁₁H₁₉NO₆·C₁₂H₂₃N
- MDL Number:MFCD01861340
- Molecular Weight:442.6 g/mol
Specifications for Product Family
Specifications
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: A234758-1GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77152-974 - Supplier Number: A234758-5GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77152-976 - Supplier Number: A234758-25GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New Product
More
Specifications
Cat. No.77892-400New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



