Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Micro Fluorescence Flow Through Cell (Lightpath: 2 mm), Type 74FL

Micro Fluorescence Flow Through Cell (Lightpath: 2 mm), Type 74FL
 
Micro Fluorescence Flow Through Cell (Lightpath: 2 mm), Type 74FL

 

 

Firefly's micro fluorometer flow cell with a 2 mm lightpath.

  • Fire-fused to ensure that cuvettes are robust and durable
  • Available in different materials: UV quartz and IR quartz
  • Available in 2 mm lightpath
  • Four windows optically polished

Four sides of the optical chamber are polished.

Product Family Options

 

  • Capacity
    Internal dimensions
    External dimensions
    Lightpath
    Material
     
     
  • 30 µl
    L×W×H 2×2×7.6 mm
    L×W×H 3×3×7.6 mm
    2 mm
    UV quartz (190 - 2500 nm)
       
       
    Each
    Each - $1,421.41
    $1,421.41
    Each
       $1,421.41

     : 74FLUV2
  • 30 µl
    L×W×H 2×2×7.6 mm
    L×W×H 3×3×7.6 mm
    2 mm
    IR quartz (220 - 3500 nm)
       
       
    Each
    Each - $1,458.70
    $1,458.70
    Each
       $1,458.70

     : 74FLIR2