Macro Laser Absorption Cell with PTFE Stopper (Lightpath: 30 to 60 mm), Type 87
Firefly's laser absorption cell.
- Fire-fused to ensure that cuvettes are robust and durable
- Available in UV Quartz and IR Quartz
- Lightpaths of 30 mm and 60 mm available
- Quartz tubes (Ext.Ø×Int.Ø×L):- 6×4×20 mm
- Height is without tube
Four windows are optically polished.