Order Entry
Puerto Rico
ContactUsLinkComponent
Lighthouse Foam Unit, 0.4 Gallon
Lighthouse Foam Unit, 0.4 Gallon
Catalog # 77007-828
Supplier:  Unimed-Midwest
undefined
Lighthouse Foam Unit, 0.4 Gallon
Catalog # 77007-828
Supplier:  Unimed-Midwest
Supplier Number:  FPFU51615L

Specifications

  • Description:
    Foam unit, 0.4 gallon
  • Cat. No.:
    77007-828

Specifications

About this item

This small foaming unit is perfect for small spaces and quick disinfections. Best used in restrooms, vehicles, and other spaces that have targeted disinfection areas.

  • Comfortable grip handle with trigger for quick on/off control
  • Nozzle with 110° fan tip
  • Pressure relief valve for safety
  • Viton pump seals for chemical compatibility
  • Polypropylene tank and pump head
  • View stripe for easy visibility of product levels
  • Stability base with gripping notches

Compact unit with manual pump provides maximum portability for small jobs and spot cleaning.

Color-coding options include black or white finishes.