ContactUsLinkComponent
Lighthouse Battery Foam Unit with Stainless Steel Ball Valve, 10 Gallon
Lighthouse Battery Foam Unit with Stainless Steel Ball Valve, 10 Gallon
Catalog # 77007-820
Supplier:  Unimed-Midwest
undefined
Lighthouse Battery Foam Unit with Stainless Steel Ball Valve, 10 Gallon
Catalog # 77007-820
Supplier:  Unimed-Midwest
Supplier Number:  FPFU516SBV
Specifications
  • Description:
    Battery foam unit with stainless steel ball valve, 10 gallon
  • Cat. No.:
    77007-820
Specifications
About this item
Made for large coverage areas, this unit holds up to 12.5 gallons of fluid and boasts an output distance of 15 to 20 feet.
  • Draws from pre-mixed solution
  • Battery operated
  • Up to 45 minutes of continuous use from one charge
  • Stainless ball valve with foam fan tip
  • Hinged back plate provides tool-free access to components
  • Natural color tank for easy visibility of product levels
  • Hinged lid
  • Integrated hose wraps
Compatible with Peroxigard® RTU (Ready To Use) and concentrate from lighthouse life sciences.
Color-coding options include 5 lid colors (black, blue, green, yellow, red), and 4 discharge hose colors (blue, green, yellow, red).