Twin-Line Pre-Mix Foam Unit
Catalog # 77007-808
Supplier: Unimed-Midwest
Specifications
- Description:Twin-line pre-mix foam unit
- Cat. No.:77007-808
Specifications
About this item
The FI-TL is a wall-mounted foam unit for use with pre-mixed or ready-to-use foaming chemical products.
- Draws from pre-mixed solution
- Powered by compressed air
- Compact design for easy use
- 65 degree fan tip for increased coverage
- Supports up to 3 twin-line stainless steel nozzle assemblies
Unit includes 25' (7.6 m) of coiled discharge tubing, and provides up to 100' (30.5 m) total distance between control box and nozzle(s) (distance does not need to be evenly distributed).