ContactUsLinkComponent
Hydro Satellite Sprayer Dilution System
Hydro Satellite Sprayer Dilution System
Catalog # 77007-814
Supplier:  Unimed-Midwest
undefined
Hydro Satellite Sprayer Dilution System
Catalog # 77007-814
Supplier:  Unimed-Midwest
Supplier Number:  HSMD412844
Specifications
  • Description:
    Hydro satellite sprayer dilution system
  • Environmentally Preferable:
  • Cat. No.:
    77007-814
Specifications
About this item
The satellite mobile dispensing system offers a low-cost, portable means of accurately diluting concentrated cleaning products for use in mop buckets, sinks, auto scrubbers, tanks, and other containers.
  • Easy mobility
  • Adjustable product connection
  • Handy 'on/off' trigger and convenient locking button
  • Over molded handle and built-in hanger
  • Adjustable velcro strap for container stability
  • Multiple dilution rates
Designed to fit virtually any cap, this dispensing system provides easy connections to a water source, and offers multiple dilution rates. The unit features exclusive E-gap eductor technology as well as AccuPro pressure regulation technology (patent pending).