ContactUsLinkComponent
PSA™ iFluor® 647 Imaging Kit with Goat Anti-Mouse IgG
PSA™ iFluor® 647 Imaging Kit with Goat Anti-Mouse IgG
  76484-932
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
undefined
PSA™ iFluor® 647 Imaging Kit with Goat Anti-Mouse IgG
  76484-932
 :  45290
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 - 50 times higher than the widely used tyramide (TSA) reagents
  • Size:
    100 tests
  • Cat. No.:
    76484-932
iFluor® 647 PSA kit is a much superior replacement for Alexa Fluor 647 tyramide-based kit or other spectrally similar fluorescent tyramide or TSA kits.
  • Higher efficiency
  • Improved fluorescence signal
  • Less consumption of primary antibody
  • 10 to 50X improvement over TSA reagents
In combination with our superior iFluor® dyes that have higher florescence intensity, increased photostability and enhanced water solubility, the iFluor® dye-labeled Styramide™ conjugates can generate fluorescence signal with significantly higher precision and sensitivity (more than 100 times) than standard ICC/IF/IHC.
 : Plastic vial.
 : For research use only.