ContactUsLinkComponent
Cell Explorer™ Live Cell Tracking Kit
Cell Explorer™ Live Cell Tracking Kit
Catalog # 76483-630
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Cell Explorer™ Live Cell Tracking Kit
Catalog # 76483-630
Supplier:  AAT Bioquest
Supplier Number:  22620
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Kit is designed to uniformly label live cells in blue fluorescence for the studies that require the fluorescent tag molecules retained inside cells for relatively longer time
  • Size:
    200 tests
  • Cat. No.:
    76483-630
Specifications
About this item
The kit uses a weakly fluorescent dye that carries a cell-retaining moiety.
  • Long term tracking
  • Suitable for variety of studies
  • Optimized cell labeling protocol
The labeling process is robust, requiring minimal hands-on time. It can be readily adapted for a wide variety of fluorescence platforms such as microplate assays, immunocytochemistry and flow cytometry.
Packaging: Plastic vial.
Caution: For research use only.