ContactUsLinkComponent
undefined | Accessories
Catalog # 76181-832
Supplier:  SPEX
undefined
Catalog # 76181-832
Supplier:  SPEX
Supplier Number:  2600
Specifications
  • Description:
    Cryo-Station cooling device refrigerated by liquid nitrogen
  • Environmentally Preferable:
  • Cat. No.:
    76181-832
  • For use with:
    Holds two Cryo-Blocks
Specifications
About this item
Hold temperature sensitive samples chilled prior to grinding.
  • Fill insulated jacket manually or automatically with liquid nitrogen supply
  • Quickly reach liquid nitrogen temperatures
Cryo-Station chamber offers a convenient place to keep samples that must be kept chilled prior to grinding or to preserve pesticides, RNA, or proteins for extraction. The well can hold a chilled cutting board (included) to prepare temperature sensitive samples.