- Pack type:Plastic vial
- Description:ProLite™ His-Tag Protein Gel Staining Kit "Green Fluorescence"
- Size:10 gels
- Cat. No.:76779-082
This kit requires very little hands-on time, allowing for rapid protein expression screening in a variety of gel types using an imaging equipment with a standard FITC filter.
- Green fluorescence
- Compatible with standard FITC filter
Kit provides a fast, sensitive, and highly specific fluorescent stain for visualizing His-tagged fusion proteins directly in a polyacrylamide gel following electrophoresis
It detects nanogram of His-tagged proteins directly in gels, thus eliminating the additional western blotting step.