Order Entry
Puerto Rico
ContactUsLinkComponent
2,4-Dichlorophenyl isocyanate 98%
Toxic
Health
2,4-Dichlorophenyl isocyanate 98%
Catalog # 77118-792
Supplier:  Ambeed
CAS Number:  
Toxic
Health
2,4-Dichlorophenyl isocyanate 98%
Catalog # 77118-792
Supplier:  Ambeed
Supplier Number:  A905287-1G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 g
  • Cat. No.:
    77118-792

Specifications

About this item

 :
 :188.01 g/mol
UN:2250
ADR:6.1,Ⅱ
 :GHS06
 :GHS08
CAS Number:2612-57-9
Formula:C₇H₃Cl₂NO
MDL Number:MFCD00002001