Ac-DL-Pro-OH 97%
New Product
Supplier: Ambeed
Jump to:
About this item
: for Research Use Only
Synonyms: 1-Acetylpyrrolidine-2-carboxylic acid, N-Acetyl-DL-Proline
- CAS Number:1074-79-9
- Formula:C₇H₁₁NO₃
- MDL Number:MFCD00063230
- Molecular Weight:337.38
5 Options Available Below
Product Details & Documents
: for Research Use Only
Synonyms: 1-Acetylpyrrolidine-2-carboxylic acid, N-Acetyl-DL-Proline
- CAS Number:1074-79-9
- Formula:C₇H₁₁NO₃
- MDL Number:MFCD00063230
- Molecular Weight:337.38
Product Family Options
Product Information
- SizePack typeAvailabilityPrice
- Supplier Number: A550565-10GSpecifications:New ProductNew Product
- Supplier Number: A550565-100GSpecifications:New ProductNew Product
- Supplier Number: A550565-25GSpecifications:New ProductNew Product
- Supplier Number: A550565-1GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: A550565-5GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



