Order Entry
Puerto Rico
ContactUsLinkComponent
PIP® EZ-Twist™ Polyurethane Foam Corded Ear Plugs
PIP® EZ-Twist™ Polyurethane Foam Corded Ear Plugs
Catalog # 76944-642
Supplier:  PIP
undefined
PIP® EZ-Twist™ Polyurethane Foam Corded Ear Plugs
Catalog # 76944-642
Supplier:  PIP
Supplier Number:  267-HPF610C

Specifications

  • Noise reduction rating:
    30 dB
  • Description:
    Polyurethane foam corded ear plugs
  • Cat. No.:
    76944-642

Specifications

About this item

Disposable soft polyurethane foam corded ear plugs provide NRR 30dB protection.

  • Just insert and twist
  • Fast fitting
  • Shape conforms to most ear canals
  • 76944-642 cylinder shap
  • Black PVC cord for convenience

Hybrid foam plug and unique stem enable easy insertion, with no need to roll down before fitting. Ideal for use with dirty hands. The PowerSoft™ Foam Technology features a proprietary polyurethane formulation that delivers optimal recovery time, easy insertion, and maximum all-day comfort. Shape conforms to most ear canals for maximum sound attenuation.