ContactUsLinkComponent
PIP® Bouton® Universal Fit Polycarbonate Safety Visor with Chin Cup
PIP® Bouton® Universal Fit Polycarbonate Safety Visor with Chin Cup
Catalog # 76944-634
Supplier:  PIP
undefined
PIP® Bouton® Universal Fit Polycarbonate Safety Visor with Chin Cup
Catalog # 76944-634
Supplier:  PIP
Supplier Number:  251-01-5210
Specifications
  • Description:
    Universal fit safety visor with chin cup
  • Material:
    Polycarbonate
  • Thickness:
    0.040"
  • Cat. No.:
    76944-634
Specifications
About this item
Universal fit polycarbonate safety visor with chin cup features an 0.040" thick lens.
  • Clear polycarbonate visor
  • Pre-assembled clear chin cup provides additional protection
  • Universal hole pattern will fit most headgear and hard hat adapters
  • Ships with masking on both sides to prevent scratching
Polycarbonate safety visor with aluminum binding is ideal for high impact applications. Outperforms PETG (Polyethylene Terephthalate Glycol) in a wider range of temperatures.
Certifications: Meets ANSI Z87.1+ requirements.