ContactUsLinkComponent
PIP® Recon™ Rimless Safety Glasses
PIP® Recon™ Rimless Safety Glasses
Catalog # 76944-626
Supplier:  PIP
undefined
PIP® Recon™ Rimless Safety Glasses
Catalog # 76944-626
Supplier:  PIP
Supplier Number:  250-32-0520
Specifications
  • Frame color:
    Clear
  • Description:
    Recon™ rimless safety glasses
  • Environmentally Preferable:
  • Cat. No.:
    76944-626
  • Lens tint:
    Clear
Specifications
About this item
Rimless safety glasses feature a clear temple, clear lens, and anti-scratch FogLess® 3Sixty™ coating.
  • Lightweight, rimless design provides all day comfort
  • Flexible bayonet temples contain rubber pads for a secure, non-slip fit
  • Adjustable rubber nosepiece provides a comfortable fit
  • 9 Base curve lens
Lightweight, rimless design provides an ergonomic fit, with flexible bayonet temples that contain rubber pads for an improved fit. The FogLess® 3Sixty™ enhanced hydrophobic coating resists lens fogging and provides clear vision in extreme working conditions.
Certifications: Meets ANSI Z87.1+. Meets CSA Z94.3.