Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
VWR® Probe qPCR Master Mix

VWR® Probe qPCR Master Mix

VWR® Probe qPCR Master Mix
Supplier:  VWR International
Certificates
VWR® Probe qPCR Master Mix

1 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

VWR® Probe qPCR Master Mix features a distinctive universal passive reference dye that is compatible with ROX-dependent and ROX-independent qPCR instruments.

  • For the detection and quantification of target DNA sequences using probe-based technology, such as TaqMan®.
  • Ready-to-use master mix containing all necessary components for DNA amplification and quantification, except for primers, probes, and DNA template
  • Suitable for both genomic DNA and cDNA
  • Includes Hot Start Taq DNA Polymerase for optimal performance
  • Incorporates dUTP for carryover contamination prevention
  • Supplied at a 2X concentration

VWR® Probe qPCR Master Mix is designed to facilitate rapid, sensitive, and precise detection and quantification of specific DNA and cDNA sequences via hydrolysis probes.

Caution: Store at -20°C. For research use only.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 73122-0100
  • 73122-0500
  • 73122-1000
  • 73122-2500
  • Application:
  • qPCR probe
  • qPCR probe
  • qPCR probe
  • qPCR probe

Specifications

Product Family Options

Product Information

  • No. of reactions
    Availability
    Price
  • 73122-0100
    VWR® Probe qPCR Master Mix
    VWR® Probe qPCR Master Mix
    100
    Discontinued
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    73122-0100
    Application
    qPCR probe
  • 73122-0500
    VWR® Probe qPCR Master Mix
    VWR® Probe qPCR Master Mix
    500
    Discontinued
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    73122-0500
    Application
    qPCR probe
  • 73122-1000
    VWR® Probe qPCR Master Mix
    VWR® Probe qPCR Master Mix
    1000 (2x 500)
    Log in for availability
    Log in for availability
    Each (1,000Assays)
    Each (1,000Assays) - $420.43
    $420.43
    Each (1,000Assays)
       $420.43

    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    73122-1000
    Application
    qPCR probe
  • 73122-2500
    VWR® Probe qPCR Master Mix
    VWR® Probe qPCR Master Mix
    2500 (5× 500)
    Discontinued
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    73122-2500
    Application
    qPCR probe
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.