ContactUsLinkComponent
Accessory for OV 625 Digital Disperser
Accessory for OV 625 Digital Disperser
Catalog # 76416-174
undefined
Accessory for OV 625 Digital Disperser
Catalog # 76416-174
Supplier Number:  A00000383
Specifications
  • Description:
    Double rod mini H-stand set
  • Environmentally Preferable:
  • Cat. No.:
    76416-174
Specifications
About this item
The Double rod mini H-stand set is designed for light weigh instruments and comes complete of all its parts. This stand is designed to avoid rotation of attachment and instruments thanks to the double rod and the bosshead clamp.