SW® Indigo G-065-095IN 5.9 mil Nitrile Exam Gloves, SW Sustainability Solutions
Protection that is strong enough to stand up to rigorous use while providing sensitivity and dexterity.
- Fully textured surface for wet and dry grip
- Tear-resistant beaded cuff for easy donning
- Silicone-free for critical environments
Optimal thickness for durability and tactile sensitivity.
: Gloves are packaged 100 per box, 1000 per case.
- Catalog No:
- 77008-246
- 77008-240
- Model:
- G-065-095-IN
- G-065-095-IN
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Environmentally Preferable:
- Safer Human and Environmental Health, Sustainable Packaging
- Safer Human and Environmental Health, Sustainable Packaging
- Disposable/Reusable:
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Finish:
- Finger textured
- Finger textured
- Cuff Style:
- Beaded
- Beaded
- Cuff Thickness:
- 3.9 mil
- 3.9 mil
- Finger Thickness:
- 5.9 mil
- 5.9 mil
- Palm Thickness:
- 5.0 mil
- 5.0 mil
- Tensile Strength:
- 14 Mpa
- 14 Mpa
- Ultimate Elongation:
- 400%
- 400%
- EN Norm:
- EN 420-2003, EU 2016/425
- EN 420-2003, EU 2016/425
- Environmental Certification:
- FSC Certified (BV-COC-170864), C2C Material Health Certified
- FSC Certified (BV-COC-170864), C2C Material Health Certified