Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
D-(-)-Glucamine 98%

D-(-)-Glucamine 98%

Irritant
New Product
Supplier:   Ambeed
D-(-)-Glucamine 98%
Supplier:  Ambeed
New Product
Certificates
D-(-)-Glucamine 98%

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Caution: for Research Use Only

Synonyms:  1-Amino-1-deoxy-D-glucitol, D-Glucamine, 1-Amino-1-deoxy-D-sorbitol, (-)-1-Amino-1-deoxy-D-glucitol, (2R,3R,4R,5S)-6-Aminohexane-1,2,3,4,5-pentaol
  • CAS Number:488-43-7
  • Formula:H₂NCH₂(CH(OH))₄CH₂OH
  • MDL Number:MFCD00077776
  • Molecular Weight:181.19 g/mol
Product Family Options

Product Information

  • Size
    Pack type
    Availability
    Price
  • 100 g
    Glass bottle
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $1,275.00
    $1,275.00
    Each(1/Itm/Bx)
       $1,275.00

    Supplier Number: A230328-100G
    Specifications:
    New Product
    New Product
  • 1 g
    Glass bottle
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $33.00
    $33.00
    Each (1g)
       $33.00

    Supplier Number: A230328-1G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 5 g
    Glass bottle
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $65.00
    $65.00
    Each (5g)
       $65.00

    Supplier Number: A230328-5G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 25 g
    Glass bottle
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $319.00
    $319.00
    Each (25g)
       $319.00

    Supplier Number: A230328-25G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.