Tubing Cutters, Plastic
21509-054
: Optimize
- Description:Plastic tubing cutter
- Cat. No.:21509-054
- Supplier no.:10-10-00139
These plastic tubing cutters makes clean, burr-free, right-angled cuts on polymeric tubing. Versatile and easy to use, these can be used with a variety of ODs.
- Produces defined, sharp edges when used on polymeric tubing
- Versatile and user-friendly
- For use with various ODs