ContactUsLinkComponent
OPTI-LYNX™ Holder Tube for Trap/Filter Inserts
Catalog # 83008-174
Supplier:  Optimize
undefined
OPTI-LYNX™ Holder Tube for Trap/Filter Inserts
Catalog # 83008-174
Supplier:  Optimize
Supplier Number:  11-02570-AA
Specifications
  • Description:
    OPTI-LYNX™ holder tube for trap/filter inserts
  • Cat. No.:
    83008-174
  • Supplier no.:
    11-02570-AA
Specifications
About this item
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances.
  • For use with OPTI-LYNX™ hardware
  • Installs into holder cap to make connections with trap/filter cartridges
  • Modifies an OPTI-LYNX™ holder to accept trap/filter cartridges
Installs into holder cap to make connections with OPTI-LYNX™ trap cartridges and filter inserts.