ContactUsLinkComponent
OPTI-LYNX™ Direct-Connect Guard Column Holder Kit
OPTI-LYNX™ Direct-Connect Guard Column Holder Kit
Catalog # 77100-320
Supplier:  Optimize
undefined
OPTI-LYNX™ Direct-Connect Guard Column Holder Kit
Catalog # 77100-320
Supplier:  Optimize
Supplier Number:  11-02232-AA
Specifications
  • Description:
    OPTI-LYNX™ Direct-connect guard column holder kit (OPTI-LYNX™ guard cartridges ordered separately)
  • Cat. No.:
    77100-320
  • Supplier no.:
    11-02232-AA
Specifications
About this item
The OPTI-LYNX™ Guard Column is a high-performance, cartridge-based guard system, suitable for use with any 10 to 32 analytical column due to its auto-adjusting ZDV port adapter.
  • For use with OPTI-LYNX™ Guard column cartridges
  • Quarter-turn quick-connect design rated to 6000 psi
  • Low Volume/ Low Dispersion interface for volume critical regions
  • Repeat zero-dead-volume connection to any column
  • Hardware is only plumbed once
  • Hand-tight - no tools required
OPTI-LYNX™ combines effective column protection with the convenience of quick-connect operation. Some cartridge-based guard column products offer convenience with a price: higher internal volumes. At Optimize Technologies, we refuse to compromise performance for anything. OPTI-LYNX™ utilizes low dispersion component interfaces to keep extra column volume to a minimum: connection paths have a total internal volume of less than 2 μl. You get quick-connect convenience and low dispersion, all in one package.
Delivery: Includes: Universal port adapter, holder cap, holder tube and fittings.