ContactUsLinkComponent
OPTI-LYNX™ In-Line Union Kit
OPTI-LYNX™ In-Line Union Kit
Catalog # 77100-304
Supplier:  Optimize
undefined
OPTI-LYNX™ In-Line Union Kit
Catalog # 77100-304
Supplier:  Optimize
Supplier Number:  11-02225-AB
Specifications
  • Description:
    OPTI-LYNX™ In-line union kit
  • Cat. No.:
    77100-304
  • Supplier no.:
    11-02225-AB
Specifications
About this item
If you are already using our OPTI-LYNX™ Guard Column/Pre-Column Filter Kits and want to use OPTI-LYNX™ at the bottom of the column as well, upgrade with this kit.
  • Quick-connect union solution
  • Quarter-turn design rated to 6,000 psi
  • Low Volume/ Low Dispersion interface for volume critical regions
  • True zero-dead-volume connection
  • Hardware is only plumbed once
  • Hand-tight - no tools required
It includes everything you need to equip one end of any analytical column with straight-through quick-connect operation. Can be used alone or in conjunction with other OPTI-LYNX™ products.
Delivery: Includes bayonet connector, holder cap, sealing element tube, sealing element and fittings.