Orders LinkContactUsLinkComponent
BioX PCR Plates
BioX PCR Plates
Supplier:   TWD Scientific
BioX PCR Plates
Supplier:  TWD Scientific
Certificates
BioX PCR Plates
8 Options Available Below
Product Details & Documents
PCR plates with broad thermal cycler compatibility.
  • High contrast indexing
  • Ultra thin walls
  • Crystal clear
  • Non sterile
  • Available with or without ridges also in skirtless, semi skirted or full skirted
All plate options are certified free of detectable Rnase, Dnase, DNA, and PCR inhibitors, and are tested pyrogen-free.
Product Family Options
Product Information
  • Skirt
    Availability
    Price
  • 76502-784
    PCR plate, flat rim
    PCR plate, flat rim
    Non-Skirted
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $435.89
    $435.89
    Case of 100
       $435.89
    Supplier Number: BX60-3400
  • 76502-786
    PCR plate, flat rim
    PCR plate, flat rim
    Non-Skirted
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $435.89
    $435.89
    Case of 100
       $435.89
    Supplier Number: BX60-3401
  • 76502-764
    PCR plate, flat rim
    PCR plate, flat rim
    Fully skirted
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $435.89
    $435.89
    Case of 100
       $435.89
    Supplier Number: BX60-3441
  • 76502-788
    PCR plate, flat rim, ABI® compatible
    PCR plate, flat rim, ABI® compatible
    Semi-Skirted
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $435.89
    $435.89
    Case of 100
       $435.89
    Supplier Number: BX60-3420
  • 76502-790
    PCR plate, raised rim, ABI® compatible
    PCR plate, raised rim, ABI® compatible
    Semi-Skirted
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $435.89
    $435.89
    Case of 100
       $435.89
    Supplier Number: BX60-3425
  • 76502-792
    PCR plate, raised rim, Fast®, ABI 9800 and 7500 compatible
    PCR plate, raised rim, Fast®, ABI 9800 and 7500 compatible
    Semi-Skirted
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $435.89
    $435.89
    Case of 100
       $435.89
    Supplier Number: BX60-3426
  • 76502-796
    PCR plate, raised rim, LightCycler® compatible
    PCR plate, raised rim, LightCycler® compatible
    Semi-Skirted
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $435.89
    $435.89
    Case of 100
       $435.89
    Supplier Number: BX60-3455
  • 76502-794
    PCR plate, flat rim
    PCR plate, flat rim
    Semi-Skirted
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $435.89
    $435.89
    Case of 100
       $435.89
    Supplier Number: BX60-3450