To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:VHH
- Clonality:Monoclonal
- Clone:96A3F5
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Size:200 μl
- Cross Adsorption:It has no cross-reactivity with mouse, rat, rabbit, chicken, goat or human immunoglobulins
- Form:Liquid
- Storage Buffer:Supplied in PBS (pH 7.2), containing gelatin and 0.03% ProClin300
- Storage Temperature:Stored at 4 °C and it should be protected from prolonged exposure to light. Do not freeze. This product is stable for 1 year upon receipt, when handled and stored as Instructed
- Concentration:0.5 mg/ml
- Immunogen:Llama VHH antibody
- Cat. No.:77007-498
- Packaging:Vial
Specifications
About this item
A Camelid VHH Antibody (also called single-domain antibody, sdAb or Nanobody by Ablynx) is a peptide chain of about 110 amino acids long, comprising one variable domain of a heavy-chain antibody. Like a whole antibody, it is able to bind selectively to a specific antigen. VHH antibodies allow a broad range of applications in biotechnical as well as therapeutic use due to their small size, simple production and high affinity.
- MonoRab™ technology guarantees high affinity: Kd ~10 to 11 M
- No cross reactivity with other species
- Specific to camelid IgG2 and IgG3
- Recognize conformational epitopes of the VHH domain
- Recognize both camelid and humanized VHHs
GenScript has developed a series of rabbit monoclonal (MonoRab) anti-VHH antibodies to facilitate your research of VHHs. Our anti-VHH antibodies recognize conformational epitopes of the VHH domain of camelid lgG2 and lgG3 subclasses. It is not necessary to add a tag to VHH for detection anymore.
Type:
Antigen: VHH
Clonality: Monoclonal
Clone: 96A3F5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: