Specifications
- Antigen Name:AAV
- Clonality:Monoclonal
- Clone:5G4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western Blot:Yes
- Size:40 μg
- Cross Adsorption:No
- Form:Lyophilized
- Storage Buffer:Lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
- Storage Temperature:It is stable up to 1 year at −20 °C from date of receipt. The reconstituted antibody can be stored for 2 - 3 weeks at 2...8 °C. For long term storage, aliquot and store at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml.
- Immunogen:AAVrh.1 Virus-Like Particles (VLPs)
- Tested Applications:Dot blot
- Cat. No.:77007-506
- Packaging:Vial
Specifications
About this item
Adeno-associated virus (AAV) is a single-stranded DNA virus that commonly infects humans without pathology. Therefore, AAV can be engineered to deliver DNA to target cells as a vector for gene therapy. There are several serotypes including AAV type 1, 2, 5, 6, 8, 9, DJ and rh.1. Among them, AAV2 is the best characterized and most commonly used. Cap gene of AAV gives rise to three capsid proteins, VP1, VP2 and VP3, with molecular weight of 87, 72 and 62 kDa, respectively. These capsid proteins assemble into a near-spherical protein shell of 60 subunits for AAV.
- High specificity
- High affinity
- High stability
GenScript’s AAV antibody is a useful tool for evaluation of virus expression level, quantification of virus titer, identification of capsid proteins in AAV production, and analysis of AAV immunogenicity.
Type:
Antigen: AAV
Clonality: Monoclonal
Clone: 5G4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: