Specifications
- Antigen Name:Panitumumab
- Clonality:Monoclonal
- Clone:2E7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Size:40 μg
- Cross Adsorption:No
- Form:Lyophilized
- Storage Buffer:This product is lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
- Storage Temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2...8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml.
- Immunogen:Panitumumab
- Cat. No.:77007-648
- Packaging:Vial
Specifications
About this item
Panitumumab with trade name Vectibix, is an FDA-approved drug for the treatment of patients with EGFR-expressing metastatic colorectal carcinoma with non-mutated (wild-type) KRAS after failure of fluoropyrimidine-, oxaliplatin- and irinotecan-containing chemotherapy regimens. It is a human monoclonal antibody. Panitumumab binds to the human EGF receptor (EGFR), preventing binding of activating ligands, such as the EGF and transforming growth factor-alpha. Anti-Panitumumab Antibody (2E7), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with Panitumumab.
- High specificity
- High affinity
- High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
Type:
Antigen: Panitumumab
Clonality: Monoclonal
Clone: 2E7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: