- Clonality:Monoclonal
- Clone:2C4
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Multiple
- Size:40 μg
- Cross Adsorption:No
- Form:Lyophilized
- Storage Buffer:Lyophilized with PBS, pH 7.2, contains 1% BSA and 0.02% sodium azide
- Storage Temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Immunogen:AAV2 capsids
- Cat. No.:77007-596
- Packaging:Vial
Adeno-associated virus type 2 (AAV2) is a valuable vector for gene therapy. It is one member of the human parvovirus family which depends on co-infection with helper viruses such as adenovirus or herpes virus for efficient reproduction. The AAV2 capsid (intact particle) comprises three types of subunits, VP1, VP2, and VP3, a total of 60 copies in a ratio of 1:1:8 (VP1:VP2:VP3). For the application of AAV2 vector in gene therapy, the determination of total capsid titer is essential, accurate characterization and quantification of purified AAV2 particle product represent a critical step for clinical application. GenScript Anti-AAV2 (intact particle) Antibody (2C4), mAb, Mouse is produced from cell culture in vitro under conditions free from animal-derived components.
- High specificity
- High affinity
- High stability
GenScript's AAV antibody is a useful tool for evaluation of virus expression level, quantification of virus titer, identification of capsid proteins in AAV production, and analysis of AAV immunogenicity.
Type:
Antigen:
Clonality: Monoclonal
Clone: 2C4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Multiple